| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10377535 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10377536 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1448952_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1418100_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10377534 |
- |
- |
| Mouse Genome 430 2.0 Array |
1418100_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1448952_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1448952_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1418100_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
80515_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
80515_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1100007705.mm.1 |
NR_027827.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007705.mm.1 |
NR_027827.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007705.mm.1 |
TC1100000909.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007707.mm.1 |
NR_027827.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007707.mm.1 |
NR_027827.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007707.mm.1 |
TC1100000909.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007708.mm.1 |
NR_027827.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007708.mm.1 |
NR_027827.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007708.mm.1 |
TC1100000909.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007706.mm.1 |
NR_027827.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007706.mm.1 |
NR_027827.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007706.mm.1 |
TC1100000909.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007704.mm.1 |
NR_027827.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007704.mm.1 |
NR_027827.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007704.mm.1 |
TC1100000909.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1448952_at |
- |
- |
| Mouse Expression Set 430 |
1418100_at |
- |
- |
| Mouse Expression Set 430 |
1448952_PM_at |
- |
- |
| Mouse Expression Set 430 |
1418100_PM_at |
- |
- |
| Murine Genome U74v2 Set |
161553_i_at |
- |
- |
| Murine Genome U74v2 Set |
160905_s_at |
- |
- |
| Murine Genome U74v2 Set |
161553_PM_i_at |
- |
- |
| Murine Genome U74v2 Set |
160905_PM_s_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1418100_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1448952_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1418100_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1448952_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1448952_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1418100_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1448952_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1418100_at |
- |
- |