| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700018665.mm.1 |
NM_001163763.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018665.mm.1 |
NM_001163763.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018665.mm.1 |
NM_001163764.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018665.mm.1 |
NM_001163764.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018665.mm.1 |
NM_025674.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018665.mm.1 |
NM_025674.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018665.mm.1 |
TC1700001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700018668.mm.1 |
NM_001163763.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018668.mm.1 |
NM_001163763.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018668.mm.1 |
NM_025674.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018668.mm.1 |
NM_025674.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700018668.mm.1 |
TC1700001969.mm.1 |
MTA TC |