Assay Details
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163763.1 | NP_001157235.1 | 3 - 4 | 1321 | 92 | |
| NM_001163764.1 | NP_001157236.1 | 3 - 4 | 1164 | 92 | ||
| NM_025674.2 | NP_079950.2 | 3 - 4 | 1212 | 92 | ||
| GenBank mRNA | AK004231.1 | - | 1 - 2 | 396 | 92 | |
| AK077414.1 | - | 3 - 4 | 1200 | 92 | ||
| AK078697.1 | - | 3 - 4 | 1284 | 92 | ||
| AK088479.1 | - | 3 - 4 | 1176 | 92 | ||
| BC004617.1 | - | 3 - 4 | 1142 | 3585301 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159488 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152035 | BC004617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152035 | AK088479.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71676 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23558 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143364 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152035 | BC004617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152035 | AK088479.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P505172 | BC004617.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159477 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143364 | - | - |
| Whole Mouse Genome G4122A | A_51_P505172 | BC004617.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114442 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13324599 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13218247 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13218255 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13362869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13362867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13324594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13324593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13362871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13475680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13324604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13475674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13373154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159464 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450520 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423809_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443795_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450519 | - | - |
| Mouse Genome 430 2.0 Array | 1443795_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423809_at | - | - |
| Mouse Genome 430 2.0 Array | 1423809_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443795_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 106795_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 106795_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700018661.mm.1 | NM_001163763.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018661.mm.1 | NM_001163763.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018661.mm.1 | NM_001163764.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018661.mm.1 | NM_001163764.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018661.mm.1 | NM_025674.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018661.mm.1 | NM_025674.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018661.mm.1 | TC1700001969.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018662.mm.1 | NM_001163763.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018662.mm.1 | NM_001163763.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018662.mm.1 | NM_001163764.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018662.mm.1 | NM_001163764.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018662.mm.1 | NM_025674.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018662.mm.1 | NM_025674.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018662.mm.1 | TC1700001969.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423809_at | - | - |
| Mouse Expression Set 430 | 1423809_PM_at | - | - |
| Mouse Expression Set 430 | 1443795_PM_at | - | - |
| Mouse Expression Set 430 | 1443795_at | - | - |
| Murine Genome U74v2 Set | 102354_at | - | - |
| Murine Genome U74v2 Set | 102354_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423809_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423809_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423809_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423809_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443795_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443795_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 371577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624827 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2624827 | - | - |
| mouse-6 v1.1 (Illumina) | scl49984.4.1_114-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200152 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49984.4.1_114-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2624827 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2624827 | - | - |

Tcf19
Genomic Map