Assay Details
| Gene Symbol: Actr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025914.2 | NP_080190.1 | 4 - 5 | 439 | 92 | |
| GenBank mRNA | AK008409.1 | - | 4 - 5 | 440 | 92 | |
| AK088571.1 | - | 4 - 5 | 425 | 92 | ||
| BC062137.1 | - | 4 - 5 | 416 | 6509757 | 92 | |
More Information
Set Membership: |
|

Actr6
Genomic Map