Assay Details
| Gene Symbol: Enoph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163035.1 | NP_001156507.1 | 3 - 4 | 601 | 72 | |
| NM_026421.3 | NP_080697.2 | 3 - 4 | 601 | 72 | ||
| NR_027990.1 | 3 - 4 | 572 | 72 | |||
| GenBank mRNA | AK009954.1 | - | 3 - 4 | 584 | 72 | |
| AK079062.1 | - | 3 - 4 | 595 | 72 | ||
| AK080714.1 | - | 3 - 4 | 552 | 72 | ||
| AK082747.1 | - | 3 - 4 | 576 | 72 | ||
| AK152021.1 | - | 3 - 4 | 576 | 72 | ||
| AK159356.1 | - | 3 - 4 | 574 | 72 | ||
| AK167575.1 | - | 3 - 4 | 612 | 72 | ||
| AK168696.1 | - | 3 - 4 | 470 | 72 | ||
| BC021429.1 | - | 3 - 4 | 528 | 5030617 | 72 | |
| CX235072.1 | - | 3 - 4 | 519 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Enoph1
Genomic Map