Assay Details
| Gene Symbol: Abca14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026458.4 | NP_080734.3 | 2 - 3 | 218 | 110 | |
| GenBank mRNA | AK007156.1 | - | 2 - 3 | 179 | 110 | |
| AK015996.1 | - | 2 - 3 | 175 | 110 | ||
| AK016294.1 | - | 2 - 3 | 173 | 110 | ||
| AK029665.1 | - | 2 - 3 | 222 | 110 | ||
| AY243470.1 | - | 2 - 3 | 218 | 110 | ||
| BC116805.1 | - | 2 - 3 | 149 | 110 | ||
| BC116809.1 | - | 2 - 3 | 149 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca14
Genomic Map