Assay Details
| Gene Symbol: Ccny | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026484.3 | NP_080760.2 | 3 - 4 | 746 | 115 | |
| GenBank mRNA | AK017493.1 | - | 3 - 4 | 578 | 115 | |
| AK028696.1 | - | 3 - 4 | 747 | 115 | ||
| AK076556.1 | - | 3 - 4 | 276 | 115 | ||
| BC023321.1 | - | 4 - 5 | 664 | 3995040 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccny
Genomic Map