Assay Details
| Gene Symbol: Fam166a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026624.3 | NP_080900.1 | 1 - 2 | 113 | 61 | |
| GenBank mRNA | AK016459.1 | - | 1 - 2 | 115 | 61 | |
| BC061039.1 | - | 1 - 2 | 116 | 6704633 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | BX631039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | NM_026624.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | AK016459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | BC061039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | BX631039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | NM_026624.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | AK016459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P541258 | BC061039.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5497 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | BX631039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | NM_026624.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | AK016459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | BC061039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | BX631039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | NM_026624.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | AK016459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P541258 | BC061039.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21070614 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04456865 | - | - |
| Whole Mouse Genome G4122A | A_52_P541258 | BX631039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541258 | NM_026624.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541258 | AK016459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541258 | BC061039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541258 | BX631039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541258 | NM_026624.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541258 | AK016459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P541258 | BC061039.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107912 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01885006 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01885004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21070610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01653150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01604643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01653140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01885004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01238286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21070619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01293595 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10469948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469947 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453026_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10469943 | - | - |
| Mouse Genome 430 2.0 Array | 1453026_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453026_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68222_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68222_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200002206.mm.1 | NM_026624.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002206.mm.1 | NM_026624.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002206.mm.1 | TC0200000401.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200002207.mm.1 | NM_026624.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002207.mm.1 | NM_026624.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200002207.mm.1 | TC0200000401.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453026_PM_at | - | - |
| Mouse Expression Set 430 | 1453026_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453026_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453026_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453026_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453026_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 799781 | - | - |
| ABI Mouse Genome Survey Microarray | 855301 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5420056 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420056 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2622410 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2622410 | - | - |
| mouse-6 v1.1 (Illumina) | scl21189.7.1_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420056 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21189.7.1_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234184 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234184 | - | - |

Fam166a
Genomic Map