Assay Details
| Gene Symbol: Spata3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122732.1 | NP_001116204.1 | 2 - 3 | 576 | 61 | |
| NM_027300.3 | NP_081576.2 | 2 - 3 | 579 | 61 | ||
| NM_028647.4 | NP_082923.1 | 2 - 3 | 576 | 61 | ||
| GenBank mRNA | AF399971.1 | - | 2 - 3 | 575 | 61 | |
| AK006491.1 | - | 2 - 3 | 569 | 61 | ||
| AK015193.1 | - | 2 - 3 | 574 | 61 | ||
| AK131709.1 | - | 2 - 3 | 575 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata3
Genomic Map