Assay Details
| Gene Symbol: 2810408A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027419.3 | NP_081695.2 | 4 - 5 | 946 | 95 | |
| GenBank mRNA | AK013042.1 | - | 4 - 5 | 947 | 95 | |
| AK015537.1 | - | 4 - 5 | 872 | 95 | ||
| AK051029.1 | - | 4 - 5 | 1034 | 95 | ||
| AK171674.1 | - | 4 - 5 | 993 | 95 | ||
| BC005538.1 | - | 3 - 4 | 238 | 95 | ||
| BC051546.1 | - | 4 - 5 | 907 | 5255109 | 95 | |
| BC055710.1 | - | 4 - 5 | 1051 | 95 | ||
| BC069874.1 | - | 4 - 5 | 946 | 4216824 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

2810408A11Rik
Genomic Map