| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400006402.mm.1 |
NM_001253888.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006402.mm.1 |
NM_001253888.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006402.mm.1 |
NM_001253889.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006402.mm.1 |
NM_001253889.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006402.mm.1 |
NM_027673.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006402.mm.1 |
NM_027673.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006402.mm.1 |
TC1400000789.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400006405.mm.1 |
NM_001253888.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006405.mm.1 |
NM_001253888.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006405.mm.1 |
NM_001253889.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006405.mm.1 |
NM_001253889.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006405.mm.1 |
NM_027673.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006405.mm.1 |
NM_027673.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006405.mm.1 |
TC1400000789.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400006403.mm.1 |
NM_001253888.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006403.mm.1 |
NM_001253888.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006403.mm.1 |
NM_001253889.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006403.mm.1 |
NM_001253889.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006403.mm.1 |
NM_027673.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006403.mm.1 |
NM_027673.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006403.mm.1 |
TC1400000789.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400006404.mm.1 |
NM_001253888.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006404.mm.1 |
NM_001253888.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006404.mm.1 |
NM_001253889.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006404.mm.1 |
NM_001253889.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400006404.mm.1 |
TC1400000789.mm.1 |
MTA TC |