Assay Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027890.5 | NP_082166.3 | 3 - 4 | 499 | 61 | |
| GenBank mRNA | AK004703.2 | - | 3 - 4 | 469 | 61 | |
| AK028817.1 | - | 3 - 4 | 475 | 61 | ||
| AK037059.1 | - | 3 - 4 | 481 | 61 | ||
| AK037127.1 | - | 3 - 4 | 450 | 61 | ||
| AK038944.1 | - | 3 - 4 | 500 | 61 | ||
| AK042469.1 | - | 3 - 4 | 483 | 61 | ||
| AK128959.1 | - | 3 - 4 | 471 | 61 | ||
| AK167155.1 | - | 3 - 4 | 500 | 61 | ||
| BB632717.1 | - | 3 - 4 | 501 | 61 | ||
| BC026837.1 | - | 3 - 4 | 452 | 4222578 | 61 | |
| BC066833.1 | - | 1 - 2 | 117 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038944.1, AK167155.1 transcripts.
Set Membership: |
|

Susd2
Genomic Map