Assay Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162913.1 | NP_001156385.1 | 5 - 6 | 768 | 97 | |
| NM_027890.5 | NP_082166.3 | 7 - 8 | 1128 | 97 | ||
| GenBank mRNA | AK004703.2 | - | 7 - 8 | 1098 | 97 | |
| AK033823.1 | - | 5 - 6 | 741 | 97 | ||
| AK037059.1 | - | 7 - 8 | 1110 | 97 | ||
| AK037127.1 | - | 7 - 8 | 1079 | 97 | ||
| AK042469.1 | - | 7 - 8 | 1112 | 97 | ||
| AK128959.1 | - | 7 - 8 | 1100 | 97 | ||
| AK167155.1 | - | 6 - 7 | 1310 | 97 | ||
| BC026837.1 | - | 7 - 8 | 1081 | 4222578 | 97 | |
| BC066833.1 | - | 4 - 5 | 927 | 97 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK167155.1 transcripts.
Set Membership: |
|

Susd2
Genomic Map