Assay Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162913.1 | NP_001156385.1 | 7 - 8 | 1178 | 99 | |
| NM_027890.5 | NP_082166.3 | 9 - 10 | 1538 | 99 | ||
| GenBank mRNA | AK004703.2 | - | 9 - 10 | 1508 | 99 | |
| AK028817.1 | - | 9 - 10 | 1514 | 99 | ||
| AK033823.1 | - | 7 - 8 | 1151 | 99 | ||
| AK037059.1 | - | 9 - 10 | 1520 | 99 | ||
| AK037127.1 | - | 9 - 10 | 1489 | 99 | ||
| AK038944.1 | - | 7 - 8 | 1806 | 99 | ||
| AK042469.1 | - | 9 - 10 | 1522 | 99 | ||
| AK128959.1 | - | 9 - 10 | 1510 | 99 | ||
| AK167155.1 | - | 8 - 9 | 1720 | 99 | ||
| BC026837.1 | - | 9 - 10 | 1491 | 4222578 | 99 | |
| BC066833.1 | - | 6 - 7 | 1337 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Susd2
Genomic Map