Assay Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162913.1 | NP_001156385.1 | 9 - 10 | 1589 | 82 | |
| NM_027890.5 | NP_082166.3 | 11 - 12 | 1949 | 82 | ||
| GenBank mRNA | AK004703.2 | - | 11 - 12 | 1919 | 82 | |
| AK028817.1 | - | 11 - 12 | 1925 | 82 | ||
| AK033823.1 | - | 9 - 10 | 1562 | 82 | ||
| AK037059.1 | - | 11 - 12 | 1931 | 82 | ||
| AK037127.1 | - | 11 - 12 | 1900 | 82 | ||
| AK038944.1 | - | 9 - 10 | 2217 | 82 | ||
| AK042469.1 | - | 11 - 12 | 1933 | 82 | ||
| AK128959.1 | - | 11 - 12 | 1921 | 82 | ||
| AK167155.1 | - | 10 - 11 | 2131 | 82 | ||
| BC026837.1 | - | 11 - 12 | 1902 | 4222578 | 82 | |
| BC066833.1 | - | 8 - 9 | 1748 | 82 | ||
More Information
Set Membership: |
|

Susd2
Genomic Map