Assay Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162913.1 | NP_001156385.1 | 11 - 12 | 2037 | 70 | |
| NM_027890.5 | NP_082166.3 | 13 - 14 | 2397 | 70 | ||
| GenBank mRNA | AK004703.2 | - | 13 - 14 | 2367 | 70 | |
| AK028817.1 | - | 13 - 14 | 2373 | 70 | ||
| AK033823.1 | - | 11 - 12 | 2010 | 70 | ||
| AK037059.1 | - | 13 - 14 | 2379 | 70 | ||
| AK038944.1 | - | 11 - 12 | 2665 | 70 | ||
| AK042469.1 | - | 13 - 14 | 2381 | 70 | ||
| AK128959.1 | - | 13 - 14 | 2369 | 70 | ||
| AK167155.1 | - | 12 - 13 | 2579 | 70 | ||
| BC026837.1 | - | 13 - 14 | 2350 | 4222578 | 70 | |
| BC066833.1 | - | 10 - 11 | 2196 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Susd2
Genomic Map