Assay Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027992.3 | NP_082268.2 | 4 - 5 | 592 | 84 | |
| GenBank mRNA | AK009646.1 | - | 4 - 5 | 495 | 84 | |
| BC050246.1 | - | 4 - 5 | 509 | 6313498 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem106b
Genomic Map