Assay Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028250.3 | NP_082526.2 | 6 - 7 | 1175 | 81 | |
| GenBank mRNA | AA260937.1 | - | 1 - 2 | 78 | 81 | |
| AK011781.1 | - | 6 - 7 | 1168 | 81 | ||
| AK167959.1 | - | 7 - 8 | 1115 | 81 | ||
| BC115580.1 | - | 6 - 7 | 767 | 40102241 | 81 | |
| BC115581.1 | - | 6 - 7 | 767 | 40102242 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acbd6
Genomic Map