Assay Details
| Gene Symbol: Tmem43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028766.2 | NP_083042.1 | 4 - 5 | 528 | 93 | |
| GenBank mRNA | AK004778.1 | - | 4 - 5 | 529 | 93 | |
| AK037165.1 | - | 4 - 5 | 500 | 93 | ||
| AK050173.1 | - | 4 - 5 | 567 | 93 | ||
| AK151693.1 | - | 4 - 5 | 521 | 93 | ||
| AK151766.1 | - | 4 - 5 | 546 | 93 | ||
| AK152078.1 | - | 4 - 5 | 521 | 93 | ||
| AK168559.1 | - | 4 - 5 | 536 | 93 | ||
| BC005521.1 | - | 4 - 5 | 493 | 93 | ||
| BC024933.1 | - | 4 - 5 | 515 | 3990353 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem43
Genomic Map