Assay Details
| Gene Symbol: Tmem43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028766.2 | NP_083042.1 | 11 - 12 | 1143 | 67 | |
| GenBank mRNA | AK004778.1 | - | 11 - 12 | 1144 | 67 | |
| AK037165.1 | - | 11 - 12 | 1114 | 67 | ||
| AK050173.1 | - | 11 - 12 | 1183 | 67 | ||
| AK151693.1 | - | 11 - 12 | 1136 | 67 | ||
| AK151766.1 | - | 11 - 12 | 1161 | 67 | ||
| AK152078.1 | - | 11 - 12 | 1136 | 67 | ||
| AK168559.1 | - | 11 - 12 | 1151 | 67 | ||
| BC005521.1 | - | 11 - 12 | 1109 | 67 | ||
| BC024933.1 | - | 11 - 12 | 1130 | 3990353 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639211 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136998 | AK004778.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136998 | NM_028766.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96329 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156470 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133643 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136998 | AK004778.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136998 | NM_028766.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639226 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05981081 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133643 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05981097 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156470 | - | - |
| Whole Mouse Genome G4122A | A_51_P517157 | BC024933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517157 | AK168559.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517157 | AK152078.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517157 | AK050173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517157 | AK151693.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136998 | AK004778.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136998 | NM_028766.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133643 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156470 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05740641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05740631 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05740639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05471032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05515906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05570926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05839679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05839681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05839694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05740644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05839690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05570953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05168143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05570922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05168140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24639264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05515905 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10540109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540112 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439418_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426434_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10540105 | - | - |
| Mouse Genome 430 2.0 Array | 1426434_at | - | - |
| Mouse Genome 430 2.0 Array | 1439418_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1426434_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439418_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74122_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74122_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600008511.mm.1 | NM_028766.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600008511.mm.1 | NM_028766.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600008511.mm.1 | TC0600001087.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426434_at | - | - |
| Mouse Expression Set 430 | 1439418_x_at | - | - |
| Mouse Expression Set 430 | 1426434_PM_at | - | - |
| Mouse Expression Set 430 | 1439418_PM_x_at | - | - |
| Murine Genome U74v2 Set | 98633_PM_at | - | - |
| Murine Genome U74v2 Set | 98633_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426434_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426434_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439418_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439418_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426434_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426434_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439418_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439418_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4200288 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200288 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2700505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2700505 | - | - |
| mouse-6 v1.1 (Illumina) | scl29744.12_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29744.12_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2831283 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2700505 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2831283 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2700505 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29744.12_13-S | - | - |

Tmem43
Genomic Map