Assay Details
| Gene Symbol: Cmtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028791.5 | NP_083067.1 | 4 - 5 | 614 | 81 | |
| GenBank mRNA | AK005044.2 | - | 4 - 5 | 521 | 81 | |
| AK042060.1 | - | 4 - 5 | 560 | 81 | ||
| AK043733.1 | - | 4 - 5 | 551 | 81 | ||
| AK049190.1 | - | 4 - 5 | 598 | 81 | ||
| AK080379.1 | - | 4 - 5 | 615 | 81 | ||
| AK088624.1 | - | 4 - 5 | 549 | 81 | ||
| AK150392.1 | - | 4 - 5 | 540 | 81 | ||
| AK151194.1 | - | 4 - 5 | 540 | 81 | ||
| AK152276.1 | - | 4 - 5 | 540 | 81 | ||
| AK152452.1 | - | 4 - 5 | 521 | 81 | ||
| AK152799.1 | - | 4 - 5 | 540 | 81 | ||
| AK153021.1 | - | 4 - 5 | 540 | 81 | ||
| AK153066.1 | - | 4 - 5 | 540 | 81 | ||
| AK172883.1 | - | 1 - 2 | 97 | 81 | ||
| BC024691.1 | - | 4 - 5 | 536 | 3668655 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtr1
Genomic Map