Assay Details
| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029083.2 | NP_083359.1 | 1 - 2 | 73 | 73 | |
| GenBank mRNA | AK017926.1 | - | 1 - 2 | 73 | 73 | |
| AK081046.1 | - | 1 - 2 | 137 | 73 | ||
| BC131992.1 | - | 1 - 2 | 72 | 40130269 | 73 | |
| BC132645.1 | - | 1 - 2 | 72 | 40130922 | 73 | |
| BC145370.1 | - | 1 - 2 | 72 | 9053928 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK017926.1,AK081046.1,BC131992.1,BC132645.1,BC145370.1,NM_029083.2 transcripts.
Set Membership: |
|

Ddit4
Genomic Map