Assay Details
| Gene Symbol: Tmbim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029141.4 | NP_083417.1 | 6 - 7 | 886 | 57 | |
| NM_029154.1 | NP_083430.1 | 6 - 7 | 652 | 57 | ||
| GenBank mRNA | AK015666.1 | - | 6 - 7 | 886 | 57 | |
| AK015761.1 | - | 6 - 7 | 477 | 57 | ||
| AK161344.1 | - | 4 - 5 | 347 | 57 | ||
| BC115681.2 | - | 6 - 7 | 744 | 40104180 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054658 | AK015666.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054658 | AK015761.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054658 | NM_029154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054658 | NM_029141.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452212 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345645 | NM_146499.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35604 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129476 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156574 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054658 | AK015666.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054658 | AK015761.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054658 | NM_029154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054658 | NM_029141.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446334 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01029108 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05466563 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129476 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156574 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05466596 | - | - |
| Whole Mouse Genome G4122A | A_51_P446934 | BC115681.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446934 | AK015666.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446934 | AK161344.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446934 | NM_029154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446934 | NM_029141.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116191 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01029108 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156574 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129476 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04807517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04246125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04932530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04180556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04724128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04288480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04246126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04206784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04724123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04206783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04528599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04212704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04180554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23446369 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10519387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519381 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10519388 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432401_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429353_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10519380 | - | - |
| Mouse Genome 430 2.0 Array | 1432401_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432401_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429353_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429353_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75010_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75010_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500000148.mm.1 | NM_029141.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500000148.mm.1 | NM_029141.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500000148.mm.1 | NM_029154.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500000148.mm.1 | NM_029154.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500000148.mm.1 | TC0500000017.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1432401_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429353_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429353_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432401_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 605961 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2350133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002350133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2604216 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2604216 | - | - |
| mouse-6 v1.1 (Illumina) | scl26891.11.1_145-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2350133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26891.11.1_145-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2604216 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2604216 | - | - |

Tmbim7
Genomic Map