Assay Details
| Gene Symbol: 4930550C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029247.3 | NP_083523.1 | 4 - 5 | 273 | 112 | |
| GenBank mRNA | AK016083.1 | - | 4 - 5 | 274 | 112 | |
| AK153618.1 | - | 2 - 3 | 170 | 112 | ||
| BC115638.1 | - | 2 - 3 | 174 | 40102982 | 112 | |
| BC115639.1 | - | 4 - 5 | 261 | 40102983 | 112 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC115638.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400311 | BC115639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400311 | AK016083.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400311 | NM_029247.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400311 | BC115639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400311 | AK016083.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400311 | NM_029247.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744016 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31622 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154842 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101149 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400311 | BC115639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400311 | AK016083.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400311 | NM_029247.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400311 | BC115639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400311 | AK016083.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400311 | NM_029247.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26520419 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154842 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06792065 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101149 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06792041 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06792076 | - | - |
| Whole Mouse Genome G4122A | A_51_P400311 | BC115639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P400311 | AK016083.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P400311 | NM_029247.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P400311 | BC115639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P400311 | AK016083.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P400311 | NM_029247.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108818 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101149 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154842 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08147058 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08147068 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08147061 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08147060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26520419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08034359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07469305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08147067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07621325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07966672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08164450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07580399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26520439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08147065 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10585348 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585354 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10585349 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429824_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10585347 | - | - |
| Mouse Genome 430 2.0 Array | 1429824_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429824_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75311_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75311_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900004569.mm.1 | TC0900000641.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900004573.mm.1 | NM_029247.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004573.mm.1 | NM_029247.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900004573.mm.1 | TC0900000641.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429824_PM_at | - | - |
| Mouse Expression Set 430 | 1429824_at | - | - |
| Murine Genome U74v2 Set | 166662_PM_at | - | - |
| Murine Genome U74v2 Set | 166662_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429824_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429824_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 360483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6510402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006510402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250814 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250814 | - | - |
| mouse-6 v1.1 (Illumina) | scl36948.11.1_155-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6510402 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36948.11.1_155-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250814 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250814 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36948.11.1_155-S | - | - |

4930550C14Rik
Genomic Map