Assay Details
| Gene Symbol: 1700001C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029285.1 | NP_083561.1 | 1 - 2 | 109 | 85 | |
| GenBank mRNA | AK005574.1 | - | 1 - 2 | 109 | 85 | |
| AK006710.1 | - | 1 - 2 | 142 | 85 | ||
| BC060996.1 | - | 1 - 2 | 112 | 6743526 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111825 | AK005574.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111825 | NM_029285.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747846 | AK155221.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747846 | AK171048.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747846 | BC003330.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747846 | BC046788.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747846 | NM_145566.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747846 | NM_178162.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958690 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543416 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95993 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153001 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162555 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111825 | AK005574.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111825 | NM_029285.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23572623 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162555 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01064682 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153001 | - | - |
| Whole Mouse Genome G4122A | A_51_P109976 | AK005574.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P109976 | BC060996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P109976 | NM_029285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P263092 | AK005574.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P263092 | AK006710.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P263092 | BC060996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P263092 | NM_029285.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102947 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162555 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153001 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04132604 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04132594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23572655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04963058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04335022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04914731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04914728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04335018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04639370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04963063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04932888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04227465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04780331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04132596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04335034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04361085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04480196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04156757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04384681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04360068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04480195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04327921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04665253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04646518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04639369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04763719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04480201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23572627 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10520502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520503 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432513_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432525_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10520501 | - | - |
| Mouse Genome 430 2.0 Array | 1432525_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432513_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432513_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432525_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75434_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75434_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500002287.mm.1 | NM_029285.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002287.mm.1 | NM_029285.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002287.mm.1 | TC0500000292.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002288.mm.1 | NM_029285.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002288.mm.1 | NM_029285.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002288.mm.1 | TC0500000292.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432513_a_at | - | - |
| Mouse Expression Set 430 | 1432513_PM_a_at | - | - |
| Mouse Expression Set 430 | 1432525_at | - | - |
| Mouse Expression Set 430 | 1432525_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432525_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432525_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432513_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432513_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 479394 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 100460014 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260471 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260471 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626275 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217411 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2626275 | - | - |
| mouse-6 v1.1 (Illumina) | scl27979.4.1_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100460014 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260471 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27979.4.1_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260471 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100460014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3003501 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3003501 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2547667 | - | - |

1700001C02Rik
Genomic Map