Assay Details
| Gene Symbol: Ssmem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027073.1 | NP_081349.1 | 2 - 3 | 504 | 93 | |
| NM_029373.4 | NP_083649.1 | 2 - 3 | 318 | 93 | ||
| GenBank mRNA | AK006038.1 | - | 2 - 3 | 505 | 93 | |
| AK006332.1 | - | 2 - 3 | 304 | 93 | ||
| BC049755.1 | - | 2 - 3 | 453 | 6773255 | 93 | |
| BU938696.1 | - | 2 - 3 | 320 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041924 | BU938696.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041924 | NM_029373.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041924 | AK006332.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126567 | NM_027073.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126567 | AK006038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126567 | BC049755.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719464 | AK035433.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719464 | AK035905.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719464 | AK046400.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719464 | AK079699.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719464 | AK158207.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719464 | NM_001037099.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719464 | NM_146123.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470337 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89987 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174005 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126072 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126567 | NM_027073.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126567 | AK006038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126567 | BC049755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041924 | BU938696.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041924 | NM_029373.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041924 | AK006332.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24332424 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05849960 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126072 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174005 | - | - |
| Whole Mouse Genome G4122A | A_51_P445260 | NM_027073.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445260 | NM_029373.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445260 | AK006038.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445260 | AK006332.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445260 | BC049755.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114345 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126072 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174005 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05814461 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05814460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24332508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05038928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05009572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05521524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05038920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05812427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05098041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05569318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05569312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05521522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05038926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05436281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05813822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05038925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05812428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05098039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24332483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05814459 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10537013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10537012 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454010_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10537007 | - | - |
| Mouse Genome 430 2.0 Array | 1454010_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1454010_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75647_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75647_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600001923.mm.1 | NM_027073.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001923.mm.1 | NM_027073.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001923.mm.1 | NM_029373.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001923.mm.1 | NM_029373.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001923.mm.1 | TC0600000268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600001924.mm.1 | NM_027073.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001924.mm.1 | NM_027073.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001924.mm.1 | NM_029373.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001924.mm.1 | NM_029373.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001924.mm.1 | TC0600000268.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454010_a_at | - | - |
| Mouse Expression Set 430 | 1454010_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454010_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454010_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 695876 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940739 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 940739 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613687 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613687 | - | - |
| mouse-6 v1.1 (Illumina) | scl30266.5.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940739 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30266.5.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940739 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2613687 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2613687 | - | - |

Ssmem1
Genomic Map