| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200016475.mm.1 |
NM_029530.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016475.mm.1 |
NM_029530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016475.mm.1 |
TC0200002072.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200016480.mm.1 |
NM_029530.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016480.mm.1 |
NM_029530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016480.mm.1 |
TC0200002072.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200016473.mm.1 |
NM_029530.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016473.mm.1 |
NM_029530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016473.mm.1 |
TC0200002072.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200016481.mm.1 |
NM_029530.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016481.mm.1 |
NM_029530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016481.mm.1 |
TC0200002072.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200016474.mm.1 |
NM_029530.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016474.mm.1 |
NM_029530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016474.mm.1 |
TC0200002072.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200016476.mm.1 |
NM_029530.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016476.mm.1 |
NM_029530.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016476.mm.1 |
TC0200002072.mm.1 |
MTA TC |