Assay Details
| Gene Symbol: Dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029669.4 | NP_083945.1 | 1 - 2 | 119 | 91 | |
| GenBank mRNA | AK012521.1 | - | 1 - 2 | 72 | 91 | |
| AK030483.1 | - | 1 - 2 | 97 | 91 | ||
| BC026452.1 | - | 1 - 2 | 81 | 4460764 | 91 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK012521.1,AK030483.1,BC026452.1,NM_029669.4 transcripts.
Set Membership: |
|

Dnajc18
Genomic Map