Assay Details
| Gene Symbol: Dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029669.4 | NP_083945.1 | 4 - 5 | 636 | 96 | |
| GenBank mRNA | AK012521.1 | - | 4 - 5 | 589 | 96 | |
| AK030483.1 | - | 4 - 5 | 614 | 96 | ||
| BC026452.1 | - | 4 - 5 | 598 | 4460764 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc18
Genomic Map