Assay Details
| Gene Symbol: Efcab6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161628.1 | NP_001155100.1 | 11 - 12 | 1959 | 87 | |
| NM_001161629.1 | NP_001155101.1 | 9 - 10 | 1447 | 87 | ||
| NM_029946.4 | NP_084222.4 | 28 - 29 | 4066 | 87 | ||
| GenBank mRNA | AK016516.1 | - | 9 - 10 | 1552 | 87 | |
| AK019850.1 | - | 11 - 12 | 1959 | 87 | ||
| AK163363.1 | - | 6 - 7 | 1137 | 87 | ||
| AK220412.1 | - | 11 - 12 | 1878 | 87 | ||
| BC056955.1 | - | 16 - 17 | 2469 | 6812632 | 87 | |
| BC065111.1 | - | 28 - 29 | 4066 | 30536704 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab6
Genomic Map