| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900018623.mm.1 |
NM_030069.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018623.mm.1 |
NM_030069.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018623.mm.1 |
TC0900002233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900018630.mm.1 |
NM_030069.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018630.mm.1 |
NM_030069.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018630.mm.1 |
TC0900002233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900018624.mm.1 |
NM_030069.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018624.mm.1 |
NM_030069.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018624.mm.1 |
TC0900002233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900018632.mm.1 |
NM_030069.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018632.mm.1 |
NM_030069.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900018632.mm.1 |
TC0900002233.mm.1 |
MTA TC |