Assay Details
Gene Symbol: Hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134134.2 | NP_598895.2 | 9 - 10 | 2009 | 71 | |
NM_178277.1 | NP_840061.1 | 8 - 9 | 2104 | 71 | ||
GenBank mRNA | AB045322.1 | - | 9 - 10 | 2010 | 71 | |
AK041866.1 | - | 9 - 10 | 1987 | 71 | ||
AK129085.1 | - | 5 - 6 | 1012 | 71 | ||
AK132529.1 | - | 2 - 3 | 488 | 71 | ||
AK144993.1 | - | 8 - 9 | 1857 | 71 | ||
BC058089.1 | - | 6 - 7 | 1402 | 71 | ||
BC060046.1 | - | 2 - 3 | 434 | 71 | ||
BC066029.1 | - | 3 - 4 | 1240 | 6417061 | 71 | |
BC079580.1 | - | 8 - 9 | 2052 | 5702202 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
