Assay Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144528.3 | NP_653111.1 | 3 - 4 | 326 | 106 | |
| NR_027505.2 | 3 - 4 | 203 | 106 | |||
| GenBank mRNA | AK042319.1 | - | 3 - 4 | 193 | 106 | |
| AK154781.1 | - | 3 - 4 | 292 | 106 | ||
| BB870939.1 | - | 3 - 4 | 295 | 106 | ||
| BC016543.1 | - | 3 - 4 | 298 | 3158083 | 106 | |
| BU147682.1 | - | 3 - 4 | 390 | 106 | ||
| DV648665.1 | - | 3 - 4 | 221 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf126
Genomic Map