Assay Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144528.3 | NP_653111.1 | 4 - 5 | 566 | 71 | |
| NR_027505.2 | 4 - 5 | 443 | 71 | |||
| GenBank mRNA | AK042319.1 | - | 4 - 5 | 433 | 71 | |
| AK154781.1 | - | 4 - 5 | 532 | 71 | ||
| BC016543.1 | - | 4 - 5 | 538 | 3158083 | 71 | |
| BU147682.1 | - | 4 - 5 | 630 | 71 | ||
| CA321804.1 | - | 1 - 2 | 73 | 71 | ||
| DV648665.1 | - | 4 - 5 | 461 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf126
Genomic Map