Assay Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144528.3 | NP_653111.1 | 6 - 7 | 704 | 56 | |
| GenBank mRNA | AK154781.1 | - | 6 - 7 | 670 | 56 | |
| BC016543.1 | - | 6 - 7 | 676 | 3158083 | 56 | |
| CA321804.1 | - | 3 - 4 | 211 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf126
Genomic Map