Assay Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144528.3 | NP_653111.1 | 7 - 8 | 804 | 77 | |
| NR_027505.2 | 6 - 7 | 611 | 77 | |||
| GenBank mRNA | AK042319.1 | - | 6 - 7 | 601 | 77 | |
| AK154781.1 | - | 7 - 8 | 770 | 77 | ||
| BC016543.1 | - | 7 - 8 | 776 | 3158083 | 77 | |
| CA321804.1 | - | 4 - 5 | 311 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf126
Genomic Map