Assay Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144801.2 | NP_659050.2 | 3 - 4 | 368 | 139 | |
| GenBank mRNA | AK010209.1 | - | 3 - 4 | 441 | 139 | |
| AK035836.1 | - | 3 - 4 | 356 | 139 | ||
| AK039173.1 | - | 2 - 3 | 438 | 139 | ||
| AK087636.1 | - | 2 - 3 | 573 | 139 | ||
| BB666121.1 | - | 3 - 4 | 368 | 139 | ||
| BC017618.1 | - | 3 - 4 | 362 | 4923610 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003688 | BB739600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003688 | BC017618.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003688 | BE946904.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003688 | AK087636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003688 | NM_144801.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654540 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129596 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104338 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156072 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003688 | BB739600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003688 | BC017618.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003688 | BE946904.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003688 | AK087636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003688 | NM_144801.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095747 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01508564 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06184311 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06184333 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104338 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156072 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01508546 | - | - |
| Whole Mouse Genome G4122A | A_51_P360508 | BC017618.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360508 | AK087636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360508 | AK039173.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360508 | NM_144801.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360508 | AK035836.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119748 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127394 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156072 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104338 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987158 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987159 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987160 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05987154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05987158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06084722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06084720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06580780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06580779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05987155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06351914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06155239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06216456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06012736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095832 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10553147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553142 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10553144 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442690_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451304_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10553140 | - | - |
| Mouse Genome 430 2.0 Array | 1451304_at | - | - |
| Mouse Genome 430 2.0 Array | 1442690_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451304_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442690_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70209_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70209_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700007698.mm.1 | TC0700000868.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007698.mm.1 | NM_144801.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007698.mm.1 | NM_144801.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007697.mm.1 | TC0700000868.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700007697.mm.1 | NM_144801.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007697.mm.1 | NM_144801.2 | Refseq |
| Mouse Expression Set 430 | 1453973_PM_s_at | - | - |
| Mouse Expression Set 430 | 1453973_s_at | - | - |
| Mouse Expression Set 430 | 1442690_PM_at | - | - |
| Mouse Expression Set 430 | 1442690_at | - | - |
| Murine Genome U74v2 Set | 136067_PM_at | - | - |
| Murine Genome U74v2 Set | 136067_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451304_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451304_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442690_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442690_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 710601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5720465 | - | - |
| MouseWG-6 v2 Expression BeadChip | 7050133 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720465 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 630594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7050133 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720465 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2675595 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2731260 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608174 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2731260 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2608174 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2675595 | - | - |
| mouse-6 v1.1 (Illumina) | scl000198.1_21-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0076975.1_135-S | - | - |
| mouse-6 v1.1 (Illumina) | scl32660.11_337-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720465 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630594 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000198.1_21-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0076975.1_135-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32660.11_337-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630594 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720465 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2675595 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2731260 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2608174 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2803344 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2803344 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608174 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2731260 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2675595 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32660.11_337-S | - | - |

Tmem143
Genomic Map