| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900004827.mm.1 |
NM_007811.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004827.mm.1 |
NM_007811.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004827.mm.1 |
TC1900000538.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004829.mm.1 |
NM_007811.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004829.mm.1 |
NM_007811.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004829.mm.1 |
TC1900000538.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004826.mm.1 |
NM_007811.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004826.mm.1 |
NM_007811.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004826.mm.1 |
TC1900000538.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900004828.mm.1 |
NM_007811.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004828.mm.1 |
NM_007811.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900004828.mm.1 |
TC1900000538.mm.1 |
MTA TC |