Assay Details
| Gene Symbol: Dcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190451.1 | NP_001177380.1 | 4 - 5 | 836 | 88 | |
| NM_007833.5 | NP_031859.1 | 4 - 5 | 744 | 88 | ||
| GenBank mRNA | AK135287.1 | - | 4 - 5 | 728 | 88 | |
| AK145905.1 | - | 4 - 5 | 726 | 88 | ||
| AK147020.1 | - | 4 - 5 | 728 | 88 | ||
| AK161788.1 | - | 4 - 5 | 726 | 88 | ||
| AK165408.1 | - | 4 - 5 | 725 | 88 | ||
| BC060126.1 | - | 3 - 4 | 557 | 88 | ||
| BC132521.1 | - | 4 - 5 | 595 | 40130798 | 88 | |
| BC138564.1 | - | 4 - 5 | 595 | 8861584 | 88 | |
| X53929.1 | - | 4 - 5 | 753 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcn
Genomic Map