Assay Details
| Gene Symbol: Dsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007883.2 | NP_031909.2 | 2 - 3 | 272 | 63 | |
| GenBank mRNA | AB072269.1 | - | 2 - 3 | 300 | 63 | |
| BC131653.1 | - | 2 - 3 | 248 | 40110529 | 63 | |
| BC131654.1 | - | 2 - 3 | 248 | 40110531 | 63 | |
More Information
Set Membership: |
|

Dsg2
Genomic Map