Assay Details
| Gene Symbol: Etv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007960.4 | NP_031986.1 | 3 - 4 | 377 | 151 | |
| GenBank mRNA | AK035914.1 | - | 1 - 2 | 768 | 151 | |
| AK144713.1 | - | 3 - 4 | 326 | 151 | ||
| BC005645.1 | - | 3 - 4 | 305 | 3257346 | 151 | |
| BY122680.1 | - | 2 - 3 | 149 | 151 | ||
| CJ061156.1 | - | 3 - 4 | 317 | 151 | ||
| L10426.1 | - | 3 - 4 | 326 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Etv1
Genomic Map