Assay Details
| Gene Symbol: Etv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007960.4 | NP_031986.1 | 4 - 5 | 433 | 105 | |
| GenBank mRNA | AK035914.1 | - | 2 - 3 | 824 | 105 | |
| AK144713.1 | - | 4 - 5 | 382 | 105 | ||
| BC005645.1 | - | 4 - 5 | 361 | 3257346 | 105 | |
| BY122680.1 | - | 3 - 4 | 205 | 105 | ||
| CJ061156.1 | - | 4 - 5 | 373 | 105 | ||
| L10426.1 | - | 4 - 5 | 382 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Etv1
Genomic Map