Assay Details
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146346.1 | NP_001139818.1 | 4 - 5 | 579 | 52 | |
| NM_010129.2 | NP_034259.2 | 4 - 5 | 550 | 52 | ||
| GenBank mRNA | AK146939.1 | - | 4 - 5 | 582 | 52 | |
| BC001999.1 | - | 4 - 5 | 395 | 3485948 | 52 | |
| CK022835.1 | - | 4 - 5 | 549 | 52 | ||
| U87948.1 | - | 4 - 5 | 528 | 52 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095882 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | U87948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | BC001999.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | CK022835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | NM_001146346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | NM_010129.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | U87948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | BC001999.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | CK022835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | NM_001146346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P446510 | NM_010129.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917939 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146410 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | U87948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | BC001999.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | CK022835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | NM_001146346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | NM_010129.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | U87948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | BC001999.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | CK022835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | NM_001146346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P446510 | NM_010129.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25095884 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146410 | - | - |
| Whole Mouse Genome G4122A | A_51_P446510 | U87948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | BC001999.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | CK022835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | NM_001146346.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | NM_010129.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | U87948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | BC001999.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | CK022835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | NM_001146346.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P446510 | NM_010129.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105362 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106248 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935920 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06075454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935919 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06075450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25095882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25095841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06075452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06276196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06075450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06012737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06026048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06012735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06276195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25095835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626378 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10563446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563442 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417104_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10563441 | - | - |
| Mouse Genome 430 2.0 Array | 1417104_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417104_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13732_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13732_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700027516.mm.1 | NM_001146346.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027516.mm.1 | NM_001146346.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027516.mm.1 | NM_010129.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027516.mm.1 | NM_010129.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700027516.mm.1 | TC0700003028.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417104_PM_at | - | - |
| Mouse Expression Set 430 | 1417104_at | - | - |
| Murine Genome U74v2 Set | 93593_f_at | - | - |
| Murine Genome U74v2 Set | 93594_r_at | - | - |
| Murine Genome U74v2 Set | 93593_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93594_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417104_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417104_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417104_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417104_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1500722 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1500722 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1570039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001570039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231824 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248028 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231824 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248028 | - | - |
| mouse-6 v1.1 (Illumina) | scl31356.7.1_27-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00010.1_38-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500722 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1570039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31356.7.1_27-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00010.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500722 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248028 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2984744 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248028 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00010.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31356.7.1_27-S | - | - |

Emp3
Genomic Map