Assay Details
| Gene Symbol: Emb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010330.4 | NP_034460.3 | 3 - 4 | 663 | 129 | |
| GenBank mRNA | AK079570.1 | - | 3 - 4 | 605 | 129 | |
| AK088480.1 | - | 3 - 4 | 491 | 129 | ||
| AK138271.1 | - | 3 - 4 | 612 | 129 | ||
| AK148449.1 | - | 3 - 4 | 491 | 129 | ||
| AK148540.1 | - | 3 - 4 | 491 | 129 | ||
| AK150625.1 | - | 3 - 4 | 492 | 129 | ||
| AK161679.1 | - | 3 - 4 | 491 | 129 | ||
| AK166602.1 | - | 3 - 4 | 491 | 129 | ||
| AK167997.1 | - | 3 - 4 | 523 | 129 | ||
| BC014858.1 | - | 3 - 4 | 482 | 4500922 | 129 | |
| J03535.2 | - | 3 - 4 | 490 | 129 | ||
More Information
Set Membership: |
|

Emb
Genomic Map