Assay Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025602.3 | NP_001020773.1 | 3 - 4 | 642 | 97 | |
| NM_001294171.1 | NP_001281100.1 | 3 - 4 | 475 | 97 | ||
| NM_010743.3 | NP_034873.2 | 3 - 4 | 642 | 97 | ||
| GenBank mRNA | AK033417.1 | - | 3 - 4 | 495 | 97 | |
| AK145118.1 | - | 3 - 4 | 496 | 97 | ||
| BC051445.1 | - | 3 - 4 | 452 | 30073421 | 97 | |
| D13695.1 | - | 2 - 3 | 334 | 97 | ||
| M24843.1 | - | 3 - 4 | 483 | 97 | ||
| Y07519.1 | - | 3 - 4 | 486 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1rl1
Genomic Map