| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_001163029.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_001163029.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_001163030.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_001163030.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_001163031.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_001163031.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_011925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
NM_011925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019504.mm.1 |
TC0800002558.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_001163029.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_001163029.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_001163030.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_001163030.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_001163031.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_001163031.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_011925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
NM_011925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019499.mm.1 |
TC0800002558.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_001163029.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_001163029.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_001163030.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_001163030.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_001163031.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_001163031.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_011925.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
NM_011925.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019501.mm.1 |
TC0800002558.mm.1 |
MTA TC |