Assay Details
| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013505.3 | NP_038533.1 | 2 - 3 | 410 | 93 | |
| GenBank mRNA | AK028604.1 | - | 2 - 3 | 415 | 93 | |
| AK028636.1 | - | 2 - 3 | 411 | 93 | ||
| AK030333.1 | - | 2 - 3 | 411 | 93 | ||
| AK048547.1 | - | 2 - 3 | 411 | 93 | ||
| AK172168.1 | - | 2 - 3 | 245 | 93 | ||
| BC057867.1 | - | 2 - 3 | 190 | 5327748 | 93 | |
| L33779.1 | - | 2 - 3 | 209 | 93 | ||
More Information
Set Membership: |
|

Dsc2
Genomic Map