Assay Details
| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013505.3 | NP_038533.1 | 4 - 5 | 720 | 113 | |
| GenBank mRNA | AK028604.1 | - | 5 - 6 | 760 | 113 | |
| AK028636.1 | - | 4 - 5 | 721 | 113 | ||
| AK030333.1 | - | 4 - 5 | 721 | 113 | ||
| AK048547.1 | - | 4 - 5 | 721 | 113 | ||
| AK172168.1 | - | 4 - 5 | 555 | 113 | ||
| BC057867.1 | - | 4 - 5 | 500 | 5327748 | 113 | |
| L33779.1 | - | 4 - 5 | 519 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dsc2
Genomic Map