Assay Details
| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013505.3 | NP_038533.1 | 13 - 14 | 2378 | 74 | |
| GenBank mRNA | AK028604.1 | - | 14 - 15 | 2418 | 74 | |
| AK028636.1 | - | 13 - 14 | 2379 | 74 | ||
| AK030333.1 | - | 13 - 14 | 2379 | 74 | ||
| AK048547.1 | - | 13 - 14 | 2379 | 74 | ||
| AK078887.1 | - | 1 - 2 | 69 | 74 | ||
| AK172168.1 | - | 13 - 14 | 2213 | 74 | ||
| BC004663.1 | - | 7 - 8 | 1265 | 74 | ||
| BC057867.1 | - | 13 - 14 | 2158 | 5327748 | 74 | |
| L33779.1 | - | 13 - 14 | 2177 | 74 | ||
More Information
Set Membership: |
|

Dsc2
Genomic Map