Assay Details
| Gene Symbol: Gtpbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145979.1 | NP_001139451.1 | 6 - 7 | 956 | 59 | |
| NM_019581.3 | NP_062527.2 | 6 - 7 | 991 | 59 | ||
| GenBank mRNA | AB024573.1 | - | 4 - 5 | 627 | 59 | |
| AF168991.1 | - | 6 - 7 | 896 | 59 | ||
| AK087511.1 | - | 6 - 7 | 862 | 59 | ||
| AK146415.1 | - | 6 - 7 | 941 | 59 | ||
| AK148020.1 | - | 6 - 7 | 912 | 59 | ||
| BC049089.1 | - | 6 - 7 | 952 | 5720710 | 59 | |
| BC051958.1 | - | 6 - 7 | 952 | 5720710 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | BC049089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | BC051958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | AK146415.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | AK148020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | AF168991.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | BC049089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | BC051958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | AK146415.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | AK148020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P124089 | AF168991.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 577511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191277 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443125 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147419 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | BC049089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | BC051958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | AK146415.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | AK148020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | AF168991.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | BC049089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | BC051958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | AK146415.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | AK148020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P124089 | AF168991.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205422 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03324320 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147419 | - | - |
| Whole Mouse Genome G4122A | A_52_P2923 | BC049089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P2923 | BC051958.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P2923 | AK146415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P2923 | AK148020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P2923 | AF168991.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | BC049089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | BC051958.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | AK146415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | AK148020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | AF168991.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | BC049089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | BC051958.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | AK146415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | AK148020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P124089 | AF168991.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102923 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102447 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13360908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13319766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13319765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13360904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13490180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13360906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13218677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13218630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13373152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13389711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13218641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13218648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13490177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13218675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13472968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13490179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13319771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445454 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416690_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448437_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442305_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416691_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445442 | - | - |
| Mouse Genome 430 2.0 Array | 1442305_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448437_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448437_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416691_at | - | - |
| Mouse Genome 430 2.0 Array | 1416691_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442305_at | - | - |
| Mouse Genome 430 2.0 Array | 1416690_at | - | - |
| Mouse Genome 430 2.0 Array | 1416690_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56055_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56055_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700008355.mm.1 | NM_001145979.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008355.mm.1 | NM_001145979.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008355.mm.1 | NM_019581.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008355.mm.1 | NM_019581.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008355.mm.1 | TC1700000881.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448437_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416691_PM_at | - | - |
| Mouse Expression Set 430 | 1448437_a_at | - | - |
| Mouse Expression Set 430 | 1416690_at | - | - |
| Mouse Expression Set 430 | 1416690_PM_at | - | - |
| Mouse Expression Set 430 | 1416691_at | - | - |
| Mouse Expression Set 430 | 1442305_at | - | - |
| Mouse Expression Set 430 | 1442305_PM_at | - | - |
| Murine Genome U74v2 Set | 104144_PM_at | - | - |
| Murine Genome U74v2 Set | 104144_at | - | - |
| Murine Genome U74v2 Set | 171211_PM_at | - | - |
| Murine Genome U74v2 Set | 171211_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416691_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448437_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416690_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416691_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416690_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448437_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448437_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416690_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448437_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416691_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416691_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416690_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442305_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442305_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2760750 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102760014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760750 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600113 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2550240 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600113 | - | - |
| mouse-6 v1.1 (Illumina) | scl50683.12_301-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102760014 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760750 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50683.12_301-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102760014 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760750 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600113 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600113 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2550240 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50683.12_301-S | - | - |

Gtpbp2
Genomic Map